Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001089
Description
IRF2 Polyclonal specifically detects IRF2 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.Specifications
IRF2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
DKFZp686F0244, interferon regulatory factor 2, IRF-2 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human and Mouse. Expected from sequence similarity: Rat, Dog and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:2000 | |
NP_002190 | |
IRF2 | |
Peptide with sequence KTSDITQARVKSC corresponding to C-Terminus according to NP_002190.2. | |
0.1 mg | |
Cytokine Research | |
3660 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction