Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583159
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583159 Supplier Invitrogen™ Supplier No. PA583159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLPDIEEVKD KSIKKGNNAF RVYRMLPLSE RPSKKGKKPK TEKEDKVKHI KQEPVESSLG LSNGVSDLSP EYAVLTSTIK NEVDSTVNII VVGQSHLDSN IE Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.

IRF2 belongs to the interferon regulatory factor family. IRF2 regulates the transcription regulation important to cell proliferation as well as immune responses by binding to the upstream regulatory region of type I IFN and IFNinducible MHC class I genes (the interferon consensus sequence (ICS)) and repression of those genes. However, IRF2 also functions as a transcriptional activator of histone H4 by interactions with NF-kappa B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IRF2
Gene Accession No. P14316
Gene Alias 9830146E22Rik; AI646973; DKFZp686F0244; Interferon regulatory factor 2; IRF2; Irf-2; OTTHUMP00000218576; OTTHUMP00000218580
Gene Symbols IRF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IRF2. Recombinant protein control fragment (Product #RP-102258).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3660
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.