Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRF2 Recombinant Rabbit Monoclonal Antibody (8M2N4)
GREENER_CHOICE

Catalog No. PIMA542862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542862 Supplier Invitrogen™ Supplier No. MA542862
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa. The target is usually found in the following locations: Nucleus. Immunogen sequence: PHWRKRNIEG KQYLSNMGTR GSYLLPGMAS FVTSNKPDLQ VTIKEESNPV PYNSSWPPFQ DLPLSSSMTP ASSSSRPDRE TRASVIKKTS DITQARVKSC.

IRF2 belongs to the interferon regulatory factor family. IRF2 regulates the transcription regulation important to cell proliferation as well as immune responses by binding to the upstream regulatory region of type I IFN and IFNinducible MHC class I genes (the interferon consensus sequence (ICS)) and repression of those genes. However, IRF2 also functions as a transcriptional activator of histone H4 by interactions with NF-kappa B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 8M2N4
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IRF2
Gene Accession No. P14316
Gene Alias 9830146E22Rik; AI646973; DKFZp686F0244; Interferon regulatory factor 2; IRF2; Irf-2; OTTHUMP00000218576; OTTHUMP00000218580
Gene Symbols IRF2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 250-349 of human IRF2 (P14316).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3660
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.