Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF2BP1 Monoclonal Antibody (OTI3F4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811706

Description
IRF2BP1 acts as a transcriptional repressor. IRF2BP1 also acts as a transcriptional corepressor in a IRF2-dependent manner. This repression is not mediated at least in part by histone deacetylase activities.Specifications
IRF2BP1 | |
Monoclonal | |
Unconjugated | |
IRF2BP1 | |
interferon regulatory factor 2 binding protein 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
26145 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI3F4 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8IU81 | |
IRF2BP1 | |
Human recombit protein fragment corresponding to amino acids 457-584 of human IRF2BP1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction