Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IRF2BP1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142677
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
IRF2BP1 acts as a transcriptional repressor. IRF2BP1 also acts as a transcriptional corepressor in a IRF2-dependent manner. This repression is not mediated at least in part by histone deacetylase activities.
Specifications
| IRF2BP1 | |
| Polyclonal | |
| Unconjugated | |
| Irf2bp1 | |
| 6330414O09Rik; C530033F24; DKFZP434M154; interferon regulatory factor 2 binding protein 1; interferon regulatory factor 2-binding protein 1; IRF-2-binding protein 1; IRF2BP1; IRF-2BP1; Probable E3 ubiquitin-protein ligase IRF2BP1; Probable RING-type E3 ubiquitin transferase IRF2BP1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 26145, 272359, 308404 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8IU81, Q8R3Y8 | |
| Irf2bp1 | |
| Peptide with sequence C-QPPTQHRLVARNGE (aa 461-474). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction