Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IRF2BP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570094
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
IRF2BP1 acts as a transcriptional repressor. IRF2BP1 also acts as a transcriptional corepressor in a IRF2-dependent manner. This repression is not mediated at least in part by histone deacetylase activities.
Specifications
| IRF2BP1 | |
| Polyclonal | |
| Unconjugated | |
| Irf2bp1 | |
| 6330414O09Rik; C530033F24; DKFZP434M154; interferon regulatory factor 2 binding protein 1; interferon regulatory factor 2-binding protein 1; IRF-2-binding protein 1; IRF2BP1; IRF-2BP1; Probable E3 ubiquitin-protein ligase IRF2BP1; Probable RING-type E3 ubiquitin transferase IRF2BP1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 308404 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| Irf2bp1 | |
| synthetic peptide corresponding to a region of rat IRF2BP1. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction