Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    IRF2BP2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281995MFV500
Description
IRF2BP2 Polyclonal antibody specifically detects IRF2BP2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| IRF2BP2 | |
| Polyclonal | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| interferon regulatory factor 2 binding protein 2, IRF-2-binding protein 2 | |
| IRF2BP2 antibody was raised against an 17 amino acid synthetic peptide near the amino terminus of human IRF2BP2. The immunogen is located within amino acids 150 - 200 of IRF2BP2. Amino Acid Squence: KLEEPPELNRQSPNPRR | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified | 
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 359948 | |
| Store at 4C in the dark. | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction