Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRF2BP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584541
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584541 Supplier Invitrogen™ Supplier No. PA584541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAALILVADN AGGSHASKDA NQVHSTTRRN SNSPPSPSSM NQRRLGPREV Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 96%.

The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF2BP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IRF2BP2
Gene Accession No. Q7Z5L9
Gene Alias E130305N23Rik; interferon regulatory factor 2 binding protein 2; interferon regulatory factor 2-binding protein 2; IRF-2-binding protein 2; IRF2BP2; IRF-2BP2
Gene Symbols IRF2BP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IRF2BP2. Recombinant protein control fragment (Product #RP-107154).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 359948
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.