Learn More
IRF2BP2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303189A

Description
The recommended shelf life for this product is 1 year from date of receipt.
IRF2BP2 encodes an interferon regulatory factor-2 (IRF2)binding protein that interacts with the C-terminal transcriptionalrepression domain of IRF2. Alternative splicing results in multipletranscript variants encoding distinct isoforms.Specifications
IRF2BP2 | |
Polyclonal | |
Unconjugated | |
IRF2BP2 | |
IRF-2-binding protein 2, interferon regulatory factor 2-binding protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
359948 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q7Z5L9 | |
IRF2BP2 | |
Between 225 and 275 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.