Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3 Antibody (3F10), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104308UV
Description
IRF3 Monoclonal specifically detects IRF3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
IRF3 | |
Monoclonal | |
DyLight 350 | |
NP_001562 | |
IRF3 | |
Recombinant human IRF-3(108-166aa) purified from E. coli | |
0.1 mL | |
Immunology, Innate Immunity, Transcription Factors and Regulators | |
3661 | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, KnockDown | |
3F10 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
interferon regulatory factor 3, IRF-3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human IRF-3 mAb, clone 3F10, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human IRF3 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction