Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3 Antibody (3F10), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104308JF549
Description
IRF3 Monoclonal specifically detects IRF3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
IRF3 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IRF3 | |
Recombinant human IRF-3(108-166aa) purified from E. coli | |
0.1 mL | |
Primary | |
Anti-human IRF-3 mAb, clone 3F10, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human IRF3 protein. | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoassay | |
3F10 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
interferon regulatory factor 3, IRF-3 | |
Mouse | |
Protein G purified | |
Immunology, Innate Immunity, Transcription Factors and Regulators | |
3661 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction