Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3 Antibody (3F10), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104308MFV500
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Flow CytometrySpecifications
IRF3 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
3F10 | |
Flow Cytometry | |
interferon regulatory factor 3, IRF-3 | |
Recombinant human IRF3 (108-166aa) purified from E. coli | |
0.1 mL | |
Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
3661 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction