Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176597
Description
IRF3 Polyclonal specifically detects IRF3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IRF3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
interferon regulatory factor 3, IRF-3 | |
Rabbit | |
47 kDa | |
0.1 mg | |
Immunology, Innate Immunity, Transcription Factors and Regulators | |
3661 | |
Human, Mouse | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL | |
NP_001562 | |
IRF3 | |
Antibody was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human IRF3. The immunogen is located within amino acids 330 - 380 of IRF3. Amino Acid Squence: GESWPQDQPWTKRL | |
Affinity Purified | |
RUO | |
Primary | |
At least three isoforms of IRF3 are known to exist. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction