Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178769G
Description
IRF3 Polyclonal antibody specifically detects IRF3 in Mouse samples. It is validated for Western BlotSpecifications
IRF3 | |
Polyclonal | |
Western Blot | |
interferon regulatory factor 3, IRF-3 | |
The immunogen for this product maps to a region between residue 375 and 419 of mouse Interferon Regulatory Factor 3 using the numbering given in entry NP_058545.1 (GeneID 54131). | |
0.1 mL | |
Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
3661 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction