Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3 Antibody (OTI2G3), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271042JF549
Description
IRF3 Monoclonal specifically detects IRF3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
IRF3 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IRF3 | |
Full-length protein expressed in 293T cell transfected with human IRF3 expression vector. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2G3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
interferon regulatory factor 3, IRF-3 | |
Mouse | |
Affinity Purified | |
Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
3661.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction