Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3 Antibody (OTI2G3), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271042PCP
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
IRF3 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
OTI2G3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
interferon regulatory factor 3, IRF-3 | |
Full-length protein expressed in 293T cell transfected with human IRF3 expression vector | |
0.1 mL | |
Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
3661 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction