Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3 Antibody (PCRP-IRF3-3B2), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314106R
This item is not returnable.
View return policy
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
IRF3 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Western Blot, Immunofluorescence | |
PCRP-IRF3-3B2 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
interferon regulatory factor 3, IRF-3 | |
Recombinant full-length human IRF3 protein (Uniprot: Q14653) | |
0.1 mL | |
Immunology, Innate Immunity, Transcription Factors and Regulators | |
3661 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction