Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3 Monoclonal Antibody (OTI8G9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA809316

Description
IRF-3 is encoded by the IRF-3 gene located on chromosome 19 in humans and belongs to the Interferon regulatory transcription factor (IRF) family. IRF-3 plays a critical role in the response of the immune system to viral infection. It binds to the Interferon stimulated response element (ISRE) of its downstream target genes such as Interferon alpha and beta to activate their transcription which eventually leads to immune modulatory actions of the type I Interferon (IFN) pathway. Following phosphorylation on Serine 396, IRF-3 has been shown to affiliate with CREB binding protein (CREBBP) and translocate to the nucleus which is critical for its ability to bind to DNA and act as a transcription factor. Additionally, phosphorylation associated nuclear translocation may also protect IRF-3 from degradation by the virus machinery.Specifications
IRF3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q14653 | |
IRF3 | |
Human recombit protein fragment corresponding to amino acids 27-169 of human IRF3 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI8G9 | |
Unconjugated | |
IRF3 | |
IIAE7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3661 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction