Learn More
Invitrogen™ IRF3 Polyclonal Antibody

Description
A suggested positive control is Ramos cell lysate. PA5-20086 can be used with blocking peptide PEP-0204.
Specifications
Specifications
| Antigen | IRF3 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | IRF3 |
| Gene Accession No. | P70671, Q14653 |
| Gene Alias | C920001K05Rik; IIAE7; Interferon regulatory factor 3; IRF3; IRF-3; IRF3 (phospho S386); IRF3 (pS386); IRF3 (pSer386); IRF3 phospho; IRF3 phospho S386; IRF3 phospho Ser386; phospho IRF3 |
| Gene Symbols | IRF3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.