Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF3 Rabbit anti-Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501564997
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4997 Supplier Bethyl Laboratories, Inc Supplier No. A303384A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

IRF-3 is encoded by the IRF-3 gene located on chromosome 19 in humans and belongs to the Interferon regulatory transcription factor (IRF) family. IRF-3 plays a critical role in the response of the immune system to viral infection. It binds to the Interferon stimulated response element (ISRE) of its downstream target genes such as Interferon alpha and beta to activate their transcription which eventually leads to immune modulatory actions of the type I Interferon (IFN) pathway. Following phosphorylation on Serine 396, IRF-3 has been shown to affiliate with CREB binding protein (CREBBP) and translocate to the nucleus which is critical for its ability to bind to DNA and act as a transcription factor. Additionally, phosphorylation associated nuclear translocation may also protect IRF-3 from degradation by the virus machinery.

Specifications

Antigen IRF3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene Irf3
Gene Accession No. P70671
Gene Alias IRF-3
Gene Symbols Irf3
Host Species Rabbit
Immunogen Between 375 and 419
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54131
Target Species Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.