Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF3 Rabbit anti-Human, Mouse, Clone: 8D3Y4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $504.00
Specifications
Antigen | IRF3 |
---|---|
Clone | 8D3Y4 |
Dilution | Western Blot 1:500 - 1:2000, Knockout Validated |
Applications | Western Blot, KnockDown |
Classification | Monoclonal |
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human, Mouse samples. It is validated for Western Blot, KnockDownSpecifications
IRF3 | |
Western Blot 1:500 - 1:2000, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
interferon regulatory factor 3, IRF-3 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human IRF3 (Q14653). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
8D3Y4 | |
Western Blot, KnockDown | |
Unconjugated | |
Rabbit | |
Immunology, Innate Immunity, Transcription Factors and Regulators | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3661 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title