Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF4 Antibody (SR1923), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | IRF4 |
---|---|
Clone | SR1923 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
IRF4 Monoclonal antibody specifically detects IRF4 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
IRF4 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
interferon regulatory factor 4, IRF-4, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, NF-EM5 | |
A synthesized peptide derived from human IRF4 (Uniprot #: Q15306) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1923 | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3662 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title