Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF4 Mouse anti-Human,Mouse, PE, Clone: Q9-343, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566649
Description
IRF4 Monoclonal antibody specifically detects IRF4 in Human,Mouse samples. It is validated for Flow Cytometry.
Specifications
IRF4 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human, Mouse | |
Purified |
Flow Cytometry | |
Q9-343 | |
PE | |
25 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction