Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582031 Supplier Invitrogen™ Supplier No. PA582031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALAFSDCRLH ICLYYREILV KELTTSSPEG CRISHGHTYD ASNLDQVLFP YPEDNGQRKN IEKLLSHLER GVVLWMAPDG LYAKRLCQSR IYWDGPLALC NDRPNKLERD QTCKLFDTQQ FLSELQAFAH HGRSLPRFQV TLC Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 93%.

The IRF (interferon regulatory factor) family of transcription factors are characterized by an unique tryptophan pentad repeat DNA-binding domain. The IRFs are important in the regulation of interferons in response to infection by virus, and in the regulation of interferon-inducible genes. IRF4 is lymphocyte specific and negatively regulates toll-like-receptor (TLR) signaling that is central to the activation of innate and adaptive immune systems. A chromosomal translocation involving this gene and the IgH locus, t(6;14)(p25;q32), may be a cause of multiple myeloma. Alternatively spliced transcript variants have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IRF4
Gene Accession No. Q15306
Gene Alias AI385587; interferon regulatory factor 4; Irf4; IRF-4; LSIRF; Lymphocyte-specific interferon regulatory factor; multiple myeloma oncogene 1; MUM1; NF-EM5; PU.1 interaction partner; Sfpi1/PU.1 interaction partner; SHEP8; Spip; Transcriptional activator PIP
Gene Symbols IRF4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IRF4. Recombinant protein control fragment (Product #RP-101901).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3662
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.