Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF5 Antibody (10T1), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104307PECY7
Description
IRF5 Monoclonal specifically detects IRF5 in Human samples. It is validated for Flow Cytometry.Specifications
IRF5 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
IRF5 | |
Recombinant human IRF5 (176-240 aa) purified from E. coli | |
0.1 mL | |
Primary | |
Anti-human IRF-5 mAb, clone 10T1, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human IRF-5 (176-240 aa). | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
10T1 | |
Flow Cytometry | |
IBD14, interferon regulatory factor 5, IRF-5, SLEB10 | |
Mouse | |
Protein G purified | |
Apoptosis, Death Receptor Signaling Pathway | |
3663 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction