Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF5 Antibody (OTI1G7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271043FR
Description
IRF5 Monoclonal specifically detects IRF5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
IRF5 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
IRF5 | |
Full length recombinant protein of human IRF5 (NP_116032) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1G7 | |
Western Blot, Immunohistochemistry | |
IBD14, interferon regulatory factor 5, IRF-5, SLEB10 | |
Mouse | |
Affinity Purified | |
Apoptosis, Death Receptor Signaling Pathway | |
3663.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction