Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529742 Supplier Invitrogen™ Supplier No. PA529742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12, mouse spleen. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the interferon regulatory factor (IRF) family, a group of transcription factors with diverse roles, including virus-mediated activation of interferon, and modulation of cell growth, differentiation, apoptosis, and immune system activity. Members of the IRF family are characterized by a conserved N-terminal DNA-binding domain containing tryptophan (W) repeats. Alternative splice variants encoding different isoforms exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene IRF5
Gene Accession No. P56477, Q13568
Gene Alias AW491843; Interferon regulatory factor 5; interferon regulatory factor 5 bone marrow; IRF5; IRF-5; mirf5; OTTHUMP00000212907; SLEB10
Gene Symbols IRF5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IRF5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27056, 3663
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.