Learn More
IRF5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 105471AP150UL

Description
10547-1-AP is a rabbit polyclonal antibody raised against the C-terminal of human IRF5.
This gene encodes a member of the interferon regulatory factor (IRF) family, a group of transcription factors with diverse roles, including virus-mediated activation of interferon, and modulation of cell growth, differentiation, apoptosis, and immune system activity. Members of the IRF family are characterized by a conserved N-terminal DNA-binding domain containing tryptophan (W) repeats. Alternative splice variants encoding different isoforms exist.Specifications
IRF5 | |
Polyclonal | |
Unconjugated | |
IRF5 | |
interferon regulatory factor 5, IRF 5, IRF5, SLEB10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27056, 296953, 3663 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P56477, Q13568 | |
IRF5 | |
IRF5 Fusion Protein Ag0841 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.