Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRF6 Monoclonal Antibody (06), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA542964
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542964 100 μL
PIMA543524 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA542964 Supplier Invitrogen™ Supplier No. MA542964
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human IRF6

This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF6
Applications Immunocytochemistry
Classification Monoclonal
Clone 06
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Irf6
Gene Accession No. O14896
Gene Alias AI876454; E230028I05Rik; interferon regulatory factor 6; IRF6; IRF-6; irf6 protein; IRF-6; transcription factor; LPS; mirf6; OFC6; PIT; PPS; PPS1; transcriptional factor; VWS; VWS1; zgc:63500
Gene Symbols Irf6
Host Species Mouse
Immunogen A synthetic peptide corresponding to the C-terminus of the human IRF6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3664
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.