Learn More
Invitrogen™ IRF6 Monoclonal Antibody (OTI2A12)

Description
IRF6 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow
Specifications
Specifications
| Antigen | IRF6 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI2A12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Irf6 |
| Gene Accession No. | O14896 |
| Gene Alias | AI876454; E230028I05Rik; interferon regulatory factor 6; IRF6; IRF-6; irf6 protein; IRF-6; transcription factor; LPS; mirf6; OFC6; PIT; PPS; PPS1; transcriptional factor; VWS; VWS1; zgc:63500 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.