Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567409
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567409 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567409 Supplier Invitrogen™ Supplier No. PA567409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILVQVIPVVAR MIYEMFSGDF TRSFDSGSVR LQISTPDIKD NIVAQLKQLY RILQTQESWQ PMQPTPSMQL P Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Irf6
Gene Accession No. O14896
Gene Alias AI876454; E230028I05Rik; interferon regulatory factor 6; IRF6; IRF-6; irf6 protein; IRF-6; transcription factor; LPS; mirf6; OFC6; PIT; PPS; PPS1; transcriptional factor; VWS; VWS1; zgc:63500
Gene Symbols Irf6
Host Species Rabbit
Immunogen Recombinant Human IRF6. Recombinant protein control fragment (Product #RP-105599).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3664
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.