Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF7 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306987G
This item is not returnable.
View return policy
Description
IRF7 Polyclonal antibody specifically detects IRF7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, KnockDownSpecifications
IRF7 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, KnockDown | |
DyLight 488 | |
interferon regulatory factor 7, interferon regulatory factor-7H, IRF-7, IRF7A, IRF-7H | |
Antibody was raised against a peptide corresponding to 17 amino acids near the center of human IRF7. The immunogen is located within amino acids 200-230 of IRF7. | |
0.1 mL | |
Transcription Factors and Regulators | |
3665 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction