Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF7 Antibody (3D9), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104309H
Description
IRF7 Monoclonal specifically detects IRF7 in Human samples. It is validated for Western Blot, ELISA.Specifications
IRF7 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
IRF7 | |
Recombinant human IRF7 (1-150aa) purified from E. coli (NP_001563). | |
0.1 mL | |
Transcription Factors and Regulators | |
3665 | |
Human | |
Purified |
Western Blot, ELISA | |
3D9 | |
Western Blot, ELISA | |
interferon regulatory factor 7, interferon regulatory factor-7H, IRF-7, IRF7A, IRF-7H | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human IRF7 mAb, clone 3D9, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human IRF7 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction