Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589354 Supplier Invitrogen™ Supplier No. PA589354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ECGRSEIDEL IKEPSVDDYM GMIKRSPSPP EACRSQLLPD WWAQQPSTGV PLVTGYTTYD AHHSAFSQMV ISFYYGGKLV GQATTTCPEG CRLSLSQPGL PGTKLYGPEG LELVRFPPAD AIPSERQRQV TRKLFGHLER GVLLHSSRQG VFVKRLCQGR VFCSGNAVVC KGRPNKLERD EVVQVFDTSQ FFRELQQFYN SQGRLPDGRV VLCFGEEFPD MAPLRSKLIL VQIEQLYVRQ LAEEAGKSCG AGSVMQAPEE PPPDQVFRMF PDICASHQRS FFRENQQITV; Positive Samples: THP-1; Cellular Location: Nucleus.

IRF8 (interferon regulatory factor 8) is a transcriptional activator or repressor. It specifically binds to the upstream regulatory region of type I IFN and IFN-inducible MHC class I genes. IRF8 plays a negative regulatory role in cells of the immune system. It is involved in CD8+ dendritic cell differentiation by forming a complex with the BATF-JUNB heterodimer in immune cells. This leads to recognition of AICE sequence, an immune-specific regulatory element, followed by cooperative binding of BATF and IRF8 and activation of the genes. Mutations in the gene can result in immunodeficiencey 32A and 32B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Irf8
Gene Accession No. Q02556
Gene Alias AI893568; H-ICSBP; Icsbp; Icsbp1; IMD32A; IMD32B; interferon consensus sequence binding protein 1; Interferon consensus sequence-binding protein; Interferon regulatory factor 8; Irf8; IRF-8; Myls; transcription factor ICSBP
Gene Symbols Irf8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 137-426 of human IRF8 (NP_0021541).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3394
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.