Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF8 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA124478

Catalog No. PIPA124478


Description

Description

PA1-24478 detects Interferon regulatory factor 8 (IRF8) in human samples. PA1-24478 has been successfully used in immunohistochemistry (paraffin) and Western Blot procedures. By Western blot, PA1-24478 is specific for the ∽48 kDa Interferon regulatory factor 8 (IRF8) protein. The PA1-24478 immunogen corresponds to a synthetic peptide corresponding to a region of human IRF8. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

IRF8 (interferon regulatory factor 8) is a transcriptional activator or repressor. It specifically binds to the upstream regulatory region of type I IFN and IFN-inducible MHC class I genes. IRF8 plays a negative regulatory role in cells of the immune system. It is involved in CD8+ dendritic cell differentiation by forming a complex with the BATF-JUNB heterodimer in immune cells. This leads to recognition of AICE sequence, an immune-specific regulatory element, followed by cooperative binding of BATF and IRF8 and activation of the genes. Mutations in the gene can result in immunodeficiencey 32A and 32B.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IRF8
Polyclonal
Unconjugated
Q02556
IRF8
Synthetic peptide corresponding to a region of human IRF8.
50 μg
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
IRF8
H-ICSBP, ICSBP, ICSBP1, IRF-8
Rabbit
Affinity chromatography
RUO
3394
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.