Learn More
IRF8 Rabbit anti-Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304027A

Description
The recommended shelf life for this product is 1 year from date of receipt.
IRF8 (interferon regulatory factor 8) is a transcriptional activator or repressor. It specifically binds to the upstream regulatory region of type I IFN and IFN-inducible MHC class I genes. IRF8 plays a negative regulatory role in cells of the immune system. It is involved in CD8+ dendritic cell differentiation by forming a complex with the BATF-JUNB heterodimer in immune cells. This leads to recognition of AICE sequence, an immune-specific regulatory element, followed by cooperative binding of BATF and IRF8 and activation of the genes. Mutations in the gene can result in immunodeficiencey 32A and 32B.Specifications
IRF8 | |
Polyclonal | |
Unconjugated | |
Irf8 | |
H-ICSBP, ICSBP, ICSBP1, IRF-8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15900 | |
4° C | |
Liquid |
ChIP sequencing (ChIP-seq), Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P23611 | |
Irf8 | |
Between 200 and 250 | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.