Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF9 Antibody (PCRP-IRF9-2F8), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314228FR
This item is not returnable.
View return policy
Description
IRF9 Monoclonal antibody specifically detects IRF9 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
IRF9 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunofluorescence | |
PCRP-IRF9-2F8 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
IFN-alpha-responsive transcription factor subunit, interferon regulatory factor 9, Interferon-stimulated gene factor 3 gamma, interferon-stimulated transcription factor 3, gamma (48kD), interferon-stimulated transcription factor 3, gamma 48kDa, IRF-9, ISGF3, ISGF3 p48 subunit, ISGF3GISGF-3 gamma, p48, Transcriptional regulator ISGF3 subunit gamma | |
Recombinant full-length human IRF9 protein (Uniprot: Q00978) | |
0.1 mL | |
Signal Transduction | |
10379 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction