Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF9 Antibody (SR1836), Novus Biologicals™

Rabbit Monoclonal Antibody
$313.06 - $438.32
Specifications
| Antigen | IRF9 |
|---|---|
| Clone | SR1836 |
| Dilution | Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
Description
IRF9 Monoclonal antibody specifically detects IRF9 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| IRF9 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IFN-alpha-responsive transcription factor subunit, interferon regulatory factor 9, Interferon-stimulated gene factor 3 gamma, interferon-stimulated transcription factor 3, gamma (48kD), interferon-stimulated transcription factor 3, gamma 48kDa, IRF-9, ISGF3, ISGF3 p48 subunit, ISGF3GISGF-3 gamma, p48, Transcriptional regulator ISGF3 subunit gamma | |
| A synthesized peptide derived from human IRF9 (Uniprot #: Q00978) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1836 | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 10379 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title