Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRGC Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984010
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4010 Supplier Bioss Supplier No. BS16706R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunity-related GTPases (IRG) (also known as p47 GTPases) are a family of GTPase proteins found in vertebrates, which play critical roles in mediating innate resistance to intracellular pathogens. IRG genes have been found in a number of mammals and lower species including mice, rats, zebrafish and humans. Most of the mouse genes contain interferon-stimulated response elements which mediate transcriptional activation by IFNs. In humans, only two IRG genes have been found: human IRGC encodes a full-length IRG protein that, like the mouse homologue, is constitutively expressed in testis, while human IRGM encodes a considerably truncated protein that is constitutively expressed in cultured cells including some macrophage cell lines. As the two human genes IRGC and IRGM are not subject to IFN control, it has been suggested that the host resistance mechanism supported by IRG proteins in the mouse is lacking in humans.

Specifications

Antigen IRGC
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Irgc
Gene Accession No. Q6NXR0
Gene Alias CINEMA; F630044M05Rik; Gm1102; Gm474; Ifgge; IIGP5; immunity related GTPase cinema; Immunity-related GTPase cinema 1; immunity-related GTPase family, cinema; immunity-related GTPase family, cinema 1; interferon inducible GTPase 5; interferon inducible GTPase family member 5; interferon-gamma-inducible GTPase IFGGE protein; Interferon-inducible GTPase 5; Irgc; IRGC1; R30953_1; RGD1311107
Gene Symbols Irgc
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RGC.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56269
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.