Learn More
IRGM1 (LRG-47) Mouse anti-Mouse, Unconjugated, Clone: 1B2, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | IRGM1 (LRG-47) |
| Applications | Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 1B2 |
| Conjugate | Unconjugated |
| Gene Alias | Immunity-related GTPase family M protein 1;Interferon-inducible GTPase 3;Interferon-inducible protein 1;LPS-stimulated RAW 264.7 macrophage protein 47;LRG-47;LRG47 |
| Gene Symbols | Irgm1, Ifi1, Iigp3, Irgm |
| Host Species | Mouse |
| Immunogen | A linear peptide corresponding to 15 amino acids from the N-terminal region of mouse Immunity-related GTPase family M protein 1 (IRGM1). |
| Molecular Weight of Antigen | 46.55 kDa calculated. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.