Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRGM1 (LRG-47) Mouse anti-Mouse, Unconjugated, Clone: 1B2, MilliporeSigma™

Catalog No. MABF22642MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22642MI 25 μL
MABF22641MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22642MI Supplier MilliporeSigma Supplier No. MABF226425UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IRGM1 (LRG-47) Monoclonal antibody specifically detects IRGM1 (LRG-47) in Mouse samples. It is validated for Immunofluorescence, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRGM1 (LRG-47)
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1B2
Conjugate Unconjugated
Gene Alias Immunity-related GTPase family M protein 1;Interferon-inducible GTPase 3;Interferon-inducible protein 1;LPS-stimulated RAW 264.7 macrophage protein 47;LRG-47;LRG47
Gene Symbols Irgm1, Ifi1, Iigp3, Irgm
Host Species Mouse
Immunogen A linear peptide corresponding to 15 amino acids from the N-terminal region of mouse Immunity-related GTPase family M protein 1 (IRGM1).
Molecular Weight of Antigen 46.55 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 1B2 is a mouse monoclonal antibody that detects the long form of Immunity-related GTPase family M protein 1 (IRGM1). It targets an epitope within 15 amino acids from the N-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.