Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRGQ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565612
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565612 Supplier Invitrogen™ Supplier No. PA565612
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HYDALILVTPG APTEKDWAQV QALLLPDAPL VCVRTDGEGE DPECLGEGKM ENPKGESLKN AGGGGLENAL SKGREKCSAG SQKAGSGEGP GKAGSEGLQQ VVGMKK Highest antigen sequence identity to the following orthologs - mouse 45%, rat 43%.

IRGQ gene ontology annotations related to this gene include GTP binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRGQ
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IRGQ
Gene Accession No. Q8WZA9
Gene Alias AF322649; FKSG27; immunity related GTPase Q; immunity-related GTPase family Q protein; immunity-related GTPase family, Q; immunity-related GTPase family, Q1; immunity-related GTPase Q; Irgq; IRGQ1; RGD1309409
Gene Symbols IRGQ
Host Species Rabbit
Immunogen Recombinant Human IRGQ. Recombinant protein control fragment (Product #RP-104896).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126298
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.