Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRGQ Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS16707R

Description
IRGQ gene ontology annotations related to this gene include GTP binding.Specifications
IRGQ | |
Polyclonal | |
Unconjugated | |
IRGQ | |
FKSG27; immunity related GTPase Q; Immunity-related GTPase family Q protein; immunity-related GTPase family, Q; immunity-related GTPase family, Q1; immunity-related GTPase Q; IRGQ; IRGQ1 | |
Rabbit | |
Protein A | |
RUO | |
126298 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8WZA9 | |
IRGQ | |
KLH conjugated synthetic peptide derived from human IRGQ. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction