Iridium(IV) chloride, 99.95% (metals basis), Ir 56.5% min., Alfa Aesar™, Premion™

Manufacturer: Alfa Aesar 1218406

Quantity 5g

 View more versions of this product

Catalog No. AA1218406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10025-97-5
    Physical Form Glassy Amorphous Pieces/Powder
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula IrCl4
    Formula Weight 334.01
    MDL Number MFCD00016155
    Chemical Name or Material Iridium(IV) chloride

    Specifications

    Physical Form Glassy Amorphous Pieces/Powder
    Formula Weight 334.01
    Quantity 5g
    Health Hazard 1 H302: Harmful if swallowed.
    EINECS Number 233-048-8
    Assay Percent Range 99.95% (metals basis)
    MDL Number MFCD00016155
    CAS 10025-97-5
    Molecular Formula IrCl4
    Chemical Name or Material Iridium(IV) chloride

    H302: Harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit