Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium(IV) oxide, Premion™, 99.99% (metals basis), Ir 84.5% min

Catalog No. p-7058861
Click to view available options
Quantity:
1 g
5 g

CAS: 12030-49-8 | CH4 | 16.04 g/mol

Iridium(IV) oxide is used in the coating of anode-electrodes for industrial electrolysis in association with other rare oxides. It is also used in microelectrodes for electrophysiology research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-49-8
Molecular Formula CH4
Molecular Weight (g/mol) 16.04
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD31654066 MFCD06411993 MFCD07370731
InChI Key VNWKTOKETHGBQD-UHFFFAOYSA-N
IUPAC Name methane
SMILES C

Specifications

Color Black
Physical Form Powder
Quantity 1 g
UN Number UN1479
Solubility Information Insoluble in water.
Formula Weight 224.2
Percent Purity 99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Ir 84.5% min
Odor Odorless
Density 11.7 g/mL
Assay (metals basis)
Chemical Name or Material Iridium (IV) oxide
Show More Show Less
Hazard Category H272-H315-H319
Hazard Statement GHS H Statement
H272
May intensify fire
oxidizer.
Precautionary Statement P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 234-743-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.