Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iridium(III) acetylacetonate

Catalog No. 333352500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-335-2500MG 500 mg
33-335-225G 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-335-2500MG Supplier Sigma Aldrich Supplier No. 333352500MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iridium(III) acetylacetonate

Specifications

CAS 15635-87-7
Melting Point 269°C to 271°C
Molecular Formula C15H21IrO6
Linear Formula [CH3COCH=C(O-)CH3]3 Ir
Quantity 500 mg
MDL Number MFCD00015353
Synonym Ir(acac)3
Molecular Weight (g/mol) 489.54
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.