Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Irinotecan HydroChloride Trihydrate, 99%, PROMO, Thermo Scientific Chemicals
Topoisomerase I inhibitor
$144.60 - $278.40
Chemical Identifiers
CAS | 136572-09-3 |
---|---|
Molecular Formula | C33H39ClN4O6 |
Molecular Weight (g/mol) | 623.15 g/mol |
MDL Number | MFCD01862255 |
InChI Key | GURKHSYORGJETM-WAQYZQTGSA-N |
Synonym | irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe |
PubChem CID | 74990 |
ChEBI | CHEBI:5971 |
IUPAC Name | hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.02,11.04,9.015,20]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride |
SMILES | [H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
J62370MCPM
|
Thermo Scientific Chemicals
J62370MCPR |
100 mg |
Each for $144.60
Order in Bulk
Request bulk or custom formats |
|
|||||
J62370MDPM
|
Thermo Scientific Chemicals
J62370MDPR |
250 mg |
Each for $278.40
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Chemical Identifiers
136572-09-3 | |
623.15 g/mol | |
GURKHSYORGJETM-WAQYZQTGSA-N | |
74990 | |
hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.02,11.04,9.015,20]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride |
C33H39ClN4O6 | |
MFCD01862255 | |
irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe | |
CHEBI:5971 | |
[H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O |
Specifications
136572-09-3 | |
Yellow | |
MFCD01862255 | |
irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe | |
GURKHSYORGJETM-WAQYZQTGSA-N | |
hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.02,11.04,9.015,20]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride | |
74990 | |
99% | |
Irinotecan hydrochloride trihydrate |
256°C | |
C33H39ClN4O6 | |
14,5091 | |
Soluble in DMSO; Soluble in water with warming | |
[H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O | |
623.15 g/mol | |
CHEBI:5971 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : DW1060750
TSCA : No
Recommended Storage : Keep cold
RUO - Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title