Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Irisin/FNDC5 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP259680UV
Description
Irisin/FNDC5 Polyclonal specifically detects Irisin/FNDC5 in Human, Mouse, Camel samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Irisin/FNDC5 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FRCP2Fibronectin type III repeat-containing protein 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Camelid | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FNDC5 | |
Partial recombinant protein made to an internal region of the human Irisin/FNDC5 protein (amino acids 32-143) [UniProt Q8NAU1] | |
0.1 mL | |
Lipid and Metabolism, Neuroscience, Stem Cells | |
252995 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction