Alfa Aesar™ Iron boride, 98%

Manufacturer: Alfa Aesar™ 2295222

Quantity 100g

 View more versions of this product

Catalog No. AA2295222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12006-85-8
    Formula Weight 122.5
    MDL Number MFCD00016092
    Chemical Name or Material Iron boride

    Specifications

    Formula Weight 122.5
    Odor Odorless
    TSCA TSCA
    Mesh Size −35 Mesh
    MDL Number MFCD00016092
    CAS 12006-85-8
    Quantity 100g
    Chemical Name or Material Iron boride
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit