Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) chloride, anhydrous, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 3114118

Quantity 50g

 View more versions of this product

Catalog No. AA3114118


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7758-94-3
99.5% (Metal basis)
126.75
Iron(II) chloride, anhydrous
Powder
Cl2Fe
MFCD00011004

Specifications

7758-94-3
99.5% (Metal basis)
Off-white powder
MFCD00011004
Odorless
50g
Powder
Cl2Fe
126.75
3.16g/cm3
Off-white
Iron(II) chloride, anhydrous

Safety and Handling

SDS & Safety Documents

H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

DOTInformation : DOT Class: 8, Packing Group: III

EINECSNumber : 231-843-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit